Assay Details
| Gene Symbol: C11H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975283.1 | XP_027831084.1 | 3 - 4 | 524 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H17orf75| Gene Symbol: C11H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975283.1 | XP_027831084.1 | 3 - 4 | 524 | 69 | |
Set Membership: |
|