Assay Details
| Gene Symbol: PWWP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096168.2 | XP_014951654.2 | 1 - 2 | 661 | 103 | |
| XM_027970466.1 | XP_027826267.1 | 1 - 2 | 664 | 103 | ||
| XM_027970468.1 | XP_027826269.1 | 1 - 2 | 663 | 103 | ||
| XM_027970469.1 | XP_027826270.1 | 1 - 2 | 662 | 103 | ||
| XM_027970470.1 | XP_027826271.1 | 1 - 2 | 662 | 103 | ||
| XM_027970473.1 | XP_027826274.1 | 1 - 2 | 660 | 103 | ||
More Information
Set Membership: |
|

PWWP2A
Genomic Map