Assay Details
| Gene Symbol: LOC101120749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970511.1 | XP_027826312.1 | 7 - 8 | 1626 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120749| Gene Symbol: LOC101120749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970511.1 | XP_027826312.1 | 7 - 8 | 1626 | 106 | |
Set Membership: |
|