Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960948.1 | XP_027816749.1 | 5 - 6 | 4385 | 58 | |
| XM_027960949.1 | XP_027816750.1 | 5 - 6 | 4419 | 58 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map