Assay Details
| Gene Symbol: LOC101119694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004118.4 | XP_004004167.2 | 10 - 11 | 1431 | 84 | |
| Gene Symbol: LOC114113060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964735.1 | XP_027820536.1 | 2 - 3 | 249 | 84 | |

LOC101119694
Genomic Map