Assay Details
| Gene Symbol: LOC101119694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004118.4 | XP_004004167.2 | 12 - 13 | 1728 | 101 | |
| Gene Symbol: LOC114113060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964735.1 | XP_027820536.1 | 4 - 5 | 543 | 101 | |
More Information
Set Membership: |
|

LOC101119694
Genomic Map