Assay Details
| Gene Symbol: ST6GALNAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005615.4 | XP_004005664.2 | 3 - 4 | 446 | 106 | |
| XM_027966402.1 | XP_027822203.1 | 2 - 3 | 313 | 106 | ||
| XM_027966403.1 | XP_027822204.1 | 3 - 4 | 595 | 106 | ||
| XM_027966404.1 | XP_027822205.1 | 3 - 4 | 361 | 106 | ||
| XM_027966405.1 | XP_027822206.1 | 2 - 3 | 311 | 106 | ||
| XM_027966406.1 | XP_027822207.1 | 2 - 3 | 323 | 106 | ||
| XM_027966407.1 | XP_027822208.1 | 3 - 4 | 408 | 106 | ||
More Information
Set Membership: |
|

ST6GALNAC4
Genomic Map