Assay Details
| Gene Symbol: MTPAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014273.4 | XP_004014322.1 | 5 - 6 | 1016 | 79 | |
| XM_012188471.3 | XP_012043861.2 | 5 - 6 | 1314 | 79 | ||
More Information
Set Membership: |
|

MTPAP
Genomic Map