Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022066.4 | XP_004022115.2 | 4 - 5 | 479 | 66 | |
| XM_012106659.3 | XP_011962049.2 | 4 - 5 | 476 | 66 | ||
| XM_027963008.1 | XP_027818809.1 | 4 - 5 | 476 | 66 | ||
| XM_027963009.1 | XP_027818810.1 | 4 - 5 | 476 | 66 | ||
More Information
Set Membership: |
|

RIBC1
Genomic Map