Assay Details
| Gene Symbol: LOC101122306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980035.1 | XP_027835836.1 | 20 - 21 | 5374 | 65 | |
| XM_027980036.1 | XP_027835837.1 | 22 - 23 | 3640 | 65 | ||
| XM_027980037.1 | XP_027835838.1 | 19 - 20 | 5172 | 65 | ||
| XM_027980038.1 | XP_027835839.1 | 21 - 22 | 3439 | 65 | ||
More Information
Set Membership: |
|

LOC101122306
Genomic Map