Assay Details
| Gene Symbol: MRPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959394.1 | XP_027815195.1 | 2 - 3 | 2155 | 88 | |
| XM_027959395.1 | XP_027815196.1 | 3 - 4 | 262 | 88 | ||
| XM_027959396.1 | XP_027815197.1 | 2 - 3 | 188 | 88 | ||
More Information
Set Membership: |
|

MRPL21
Genomic Map