Assay Details
| Gene Symbol: ADRA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096166.2 | XP_014951652.2 | 1 - 2 | 1245 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRA1B| Gene Symbol: ADRA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096166.2 | XP_014951652.2 | 1 - 2 | 1245 | 74 | |
Set Membership: |
|