Assay Details
| Gene Symbol: LOC101123372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177964.3 | XP_012033354.1 | 7 - 8 | 629 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101123372| Gene Symbol: LOC101123372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177964.3 | XP_012033354.1 | 7 - 8 | 629 | 105 | |
Set Membership: |
|