Assay Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106161.3 | XP_011961551.2 | 5 - 6 | 1112 | 78 | |
| XM_027962831.1 | XP_027818632.1 | 5 - 6 | 1107 | 78 | ||
| XM_027962832.1 | XP_027818633.1 | 7 - 8 | 1122 | 78 | ||
| XM_027962833.1 | XP_027818634.1 | 7 - 8 | 1114 | 78 | ||
More Information
Set Membership: |
|

GYG2
Genomic Map