Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969482.1 | XP_027825283.1 | 13 - 14 | 1829 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADGRE3| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969482.1 | XP_027825283.1 | 13 - 14 | 1829 | 68 | |
Set Membership: |
|