Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020777.4 | XP_004020826.2 | 8 - 9 | 1328 | 102 | |
| XM_012104124.3 | XP_011959514.2 | 8 - 9 | 1331 | 102 | ||
| XM_015104083.2 | XP_014959569.1 | 8 - 9 | 1142 | 102 | ||
More Information
Set Membership: |
|

GSPT1
Genomic Map