Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002529.3 | XP_004002578.1 | 2 - 3 | 275 | 104 | |
| XM_012181932.3 | XP_012037322.1 | 2 - 3 | 288 | 104 | ||
| XM_027975424.1 | XP_027831225.1 | 1 - 2 | 177 | 104 | ||
More Information
Set Membership: |
|

S100A1
Genomic Map