Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003256.4 | XP_004003305.3 | 5 - 6 | 815 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SELENOT| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003256.4 | XP_004003305.3 | 5 - 6 | 815 | 77 | |
Set Membership: |
|