Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010484.4 | XP_004010533.1 | 3 - 4 | 472 | 78 | |
| XM_004010485.4 | XP_004010534.3 | 3 - 4 | 321 | 78 | ||
| XM_012181286.2 | XP_012036676.1 | 3 - 4 | 316 | 78 | ||
More Information
Set Membership: |
|

LRRC57
Genomic Map