Assay Details
| Gene Symbol: LOC101115252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961640.1 | XP_027817441.1 | 3 - 4 | 507 | 102 | |
| XM_027961641.1 | XP_027817442.1 | 3 - 4 | 318 | 102 | ||
| XM_027961642.1 | XP_027817443.1 | 4 - 5 | 370 | 102 | ||
| XM_027961643.1 | XP_027817444.1 | 3 - 4 | 260 | 102 | ||
| XM_027961644.1 | XP_027817445.1 | 2 - 3 | 299 | 102 | ||
More Information
Set Membership: |
|

LOC101115252
Genomic Map