Assay Details
| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006340.4 | XP_004006389.1 | 1 - 2 | 150 | 82 | |
| XM_015094662.2 | XP_014950148.1 | 1 - 2 | 116 | 82 | ||
| XM_027967308.1 | XP_027823109.1 | 1 - 2 | 107 | 82 | ||
More Information
Set Membership: |
|

BIN2
Genomic Map