Assay Details
| Gene Symbol: PPIL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011215.4 | XP_004011264.1 | 5 - 6 | 634 | 93 | |
| XM_012182617.3 | XP_012038007.1 | 5 - 6 | 634 | 93 | ||
| XM_015097351.2 | XP_014952837.1 | 5 - 6 | 592 | 93 | ||
| XM_027972383.1 | XP_027828184.1 | 5 - 6 | 634 | 93 | ||
| XM_027972384.1 | XP_027828185.1 | 5 - 6 | 634 | 93 | ||
| XM_027972386.1 | XP_027828187.1 | 5 - 6 | 634 | 93 | ||
| XM_027972387.1 | XP_027828188.1 | 5 - 6 | 634 | 93 | ||
More Information
Set Membership: |
|

PPIL6
Genomic Map