Assay Details
| Gene Symbol: DECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011847.3 | XP_004011896.3 | 2 - 3 | 281 | 61 | |
| XM_027973242.1 | XP_027829043.1 | 4 - 5 | 501 | 61 | ||
More Information
Set Membership: |
|

DECR1
Genomic Map