Assay Details
| Gene Symbol: LOC101111617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965787.1 | XP_027821588.1 | 3 - 4 | 345 | 93 | |
| XM_027965788.1 | XP_027821589.1 | 2 - 3 | 225 | 93 | ||
More Information
Set Membership: |
|

LOC101111617
Genomic Map