Assay Details
| Gene Symbol: ATF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013898.3 | XP_004013947.2 | 2 - 3 | 3169 | 79 | |
| XM_015099372.2 | XP_014954858.1 | 3 - 4 | 546 | 79 | ||
More Information
Set Membership: |
|

ATF3
Genomic Map