Assay Details
| Gene Symbol: ELAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020546.3 | XP_004020595.1 | 3 - 4 | 699 | 75 | |
| XM_004020547.3 | XP_004020596.1 | 3 - 4 | 1363 | 75 | ||
More Information
Set Membership: |
|

ELAC1
Genomic Map