Assay Details
| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956887.1 | XP_027812688.1 | 6 - 7 | 523 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM191C| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956887.1 | XP_027812688.1 | 6 - 7 | 523 | 69 | |
Set Membership: |
|