Assay Details
| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970270.1 | XP_027826071.1 | 2 - 3 | 321 | 78 | |
| XM_027970276.1 | XP_027826077.1 | 2 - 3 | 1814 | 78 | ||
More Information
Set Membership: |
|

SHISAL2A
Genomic Map