Assay Details
| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004136.4 | XP_004004185.1 | 4 - 5 | 1086 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCAF12| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004136.4 | XP_004004185.1 | 4 - 5 | 1086 | 66 | |
Set Membership: |
|