Assay Details
| Gene Symbol: ALDOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104360.2 | XP_011959750.1 | 7 - 8 | 944 | 73 | |
| XM_012104361.2 | XP_011959751.1 | 8 - 9 | 938 | 73 | ||
| XM_027962048.1 | XP_027817849.1 | 8 - 9 | 916 | 73 | ||
| XM_027962049.1 | XP_027817850.1 | 8 - 9 | 998 | 73 | ||
| XM_027962050.1 | XP_027817851.1 | 9 - 10 | 975 | 73 | ||
More Information
Set Membership: |
|

ALDOA
Genomic Map