Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022311.4 | XP_004022360.2 | 10 - 11 | 2355 | 69 | |
| XM_012107254.3 | XP_011962644.2 | 9 - 10 | 3237 | 69 | ||
| XM_012107255.3 | XP_011962645.2 | 9 - 10 | 3242 | 69 | ||
More Information
Set Membership: |
|

RBMX
Genomic Map