Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957130.1 | XP_027812931.1 | 2 - 3 | 890 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABHD17C| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957130.1 | XP_027812931.1 | 2 - 3 | 890 | 85 | |
Set Membership: |
|