Assay Details
| Gene Symbol: LOC101115509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106140.3 | XP_011961530.2 | 11 - 12 | 1553 | 87 | |
| XM_012106143.3 | XP_011961533.2 | 10 - 11 | 1457 | 87 | ||
| XM_027963315.1 | XP_027819116.1 | 11 - 12 | 1261 | 87 | ||
| XM_027963316.1 | XP_027819117.1 | 10 - 11 | 1133 | 87 | ||
More Information
Set Membership: |
|

LOC101115509
Genomic Map