Assay Details
| Gene Symbol: LOC101115509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106140.3 | XP_011961530.2 | 2 - 3 | 506 | 103 | |
| XM_012106141.3 | XP_011961531.2 | 2 - 3 | 512 | 103 | ||
| XM_012106143.3 | XP_011961533.2 | 2 - 3 | 507 | 103 | ||
| XM_027963315.1 | XP_027819116.1 | 2 - 3 | 214 | 103 | ||
More Information
Set Membership: |
|

LOC101115509
Genomic Map