Assay Details
| Gene Symbol: LOC101115509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106140.3 | XP_011961530.2 | 3 - 4 | 651 | 92 | |
| XM_012106141.3 | XP_011961531.2 | 3 - 4 | 657 | 92 | ||
| XM_012106143.3 | XP_011961533.2 | 3 - 4 | 652 | 92 | ||
| XM_027963315.1 | XP_027819116.1 | 3 - 4 | 359 | 92 | ||
More Information
Set Membership: |
|

LOC101115509
Genomic Map