Assay Details
| Gene Symbol: LOC101115509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106140.3 | XP_011961530.2 | 5 - 6 | 903 | 126 | |
| XM_012106141.3 | XP_011961531.2 | 5 - 6 | 909 | 126 | ||
| XM_012106143.3 | XP_011961533.2 | 5 - 6 | 904 | 126 | ||
| XM_027963315.1 | XP_027819116.1 | 5 - 6 | 611 | 126 | ||
| XM_027963316.1 | XP_027819117.1 | 4 - 5 | 483 | 126 | ||
More Information
Set Membership: |
|

LOC101115509
Genomic Map