Assay Details
| Gene Symbol: LOC101115509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106140.3 | XP_011961530.2 | 9 - 10 | 1372 | 85 | |
| XM_012106141.3 | XP_011961531.2 | 9 - 10 | 1378 | 85 | ||
| XM_027963315.1 | XP_027819116.1 | 9 - 10 | 1080 | 85 | ||
| XM_027963316.1 | XP_027819117.1 | 8 - 9 | 952 | 85 | ||
More Information
Set Membership: |
|

LOC101115509
Genomic Map