Assay Details
| Gene Symbol: TSPYL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973196.1 | XP_027828997.1 | 1 | 915 | 75 | |
| XM_027973197.1 | XP_027828998.1 | 1 | 915 | 75 | ||
More Information
Set Membership: |
|

TSPYL5
Genomic Map