Assay Details
| Gene Symbol: IL11RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004122.4 | XP_004004171.1 | 3 - 4 | 250 | 64 | |
| XM_012117902.3 | XP_011973292.1 | 4 - 5 | 692 | 64 | ||
| XM_012117904.3 | XP_011973294.1 | 3 - 4 | 576 | 64 | ||
| XM_027964273.1 | XP_027820074.1 | 3 - 4 | 270 | 64 | ||
More Information
Set Membership: |
|

IL11RA
Genomic Map