Assay Details
| Gene Symbol: SAMM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007040.3 | XP_004007089.1 | 9 - 10 | 1027 | 67 | |
| XM_012175752.3 | XP_012031142.1 | 9 - 10 | 998 | 67 | ||
More Information
Set Membership: |
|

SAMM50
Genomic Map