Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977671.1 | XP_027833472.1 | 5 - 6 | 876 | 82 | |
| XM_027977672.1 | XP_027833473.1 | 6 - 7 | 805 | 82 | ||
More Information
Set Membership: |
|

OSGIN1
Genomic Map