Assay Details
| Gene Symbol: LOC101116389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966004.1 | XP_027821805.1 | 4 - 5 | 501 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101116389| Gene Symbol: LOC101116389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966004.1 | XP_027821805.1 | 4 - 5 | 501 | 110 | |
Set Membership: |
|