Assay Details
| Gene Symbol: LOC101115927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003244.4 | XP_004003293.1 | 1 - 2 | 359 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115927| Gene Symbol: LOC101115927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003244.4 | XP_004003293.1 | 1 - 2 | 359 | 93 | |
Set Membership: |
|