Assay Details
| Gene Symbol: MUC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956829.1 | XP_027812630.1 | 2 - 3 | 1652 | 69 | |
| XM_027956831.1 | XP_027812632.1 | 2 - 3 | 1652 | 69 | ||
More Information
Set Membership: |
|

MUC20
Genomic Map