Assay Details
| Gene Symbol: LOC101120838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010493.4 | XP_004010542.1 | 1 | 569 | 110 | |
| XM_027972200.1 | XP_027828001.1 | 2 | 488 | 110 | ||
More Information
Set Membership: |
|

LOC101120838
Genomic Map