Assay Details
| Gene Symbol: MBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018539.4 | XP_004018588.2 | 2 - 3 | 507 | 72 | |
| XM_012100226.3 | XP_011955616.2 | 2 - 3 | 507 | 72 | ||
| XM_012100227.3 | XP_011955617.2 | 2 - 3 | 507 | 72 | ||
| XM_012100229.3 | XP_011955619.2 | 2 - 3 | 507 | 72 | ||
| XM_012100231.3 | XP_011955621.2 | 2 - 3 | 507 | 72 | ||
More Information
Set Membership: |
|

MBD4
Genomic Map