Assay Details
| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960818.1 | XP_027816619.1 | 6 - 7 | 915 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINB12| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960818.1 | XP_027816619.1 | 6 - 7 | 915 | 63 | |
Set Membership: |
|