Assay Details
| Gene Symbol: PPP1R15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190639.3 | XP_012046029.2 | 2 - 3 | 2086 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R15A| Gene Symbol: PPP1R15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190639.3 | XP_012046029.2 | 2 - 3 | 2086 | 71 | |
Set Membership: |
|