Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974521.1 | XP_027830322.1 | 3 - 4 | 871 | 84 | |
| XM_027974522.1 | XP_027830323.1 | 3 - 4 | 871 | 84 | ||
More Information
Set Membership: |
|

P3H4
Genomic Map