Assay Details
| Gene Symbol: LOC101104137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965897.1 | XP_027821698.1 | 11 - 12 | 1511 | 77 | |
| Gene Symbol: LOC114113719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965896.1 | XP_027821697.1 | 2 - 3 | 146 | 77 | |

LOC101104137
Genomic Map